Merck
Monitoring neural stem cell differentiation using PEDOT-PSS based MEA.
Biochimica et biophysica acta (2013-03-05)
Yuriko Furukawa, Akiyoshi Shimada, Koichi Kato, Hiroo Iwata, Keiichi Torimitsu
ABSTRACT
Transplantation is one potential clinical application of neural stem cells (NSCs). However, it is very difficult to monitor/control NSCs after transplantation and so provide effective treatment. Electrical measurement using a poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate) (PEDOT-PSS) modified microelectrode array (MEA) is a biocompatible, non-invasive, non-destructive approach to understanding cell conditions. This property makes continuous monitoring available for the evaluation/assessment of the development of cells such as NSCs. A PEDOT-PSS modified MEA was used to monitor electrical signals during NSC development in a culture derived from rat embryo striatum in order to understand the NSC differentiation conditions. Electrical data indicated that NSCs with nerve growth factor (NGF) generate a cultured cortical neuron-like burst pattern while a random noise pattern was measured with epidermal growth factor (EGF) at 4days in vitro (DIV) and a burst pattern was observed in both cases at 11 DIV indicating the successful monitoring of differentiation differences and developmental changes. The electrical analysis of cell activity using a PEDOT-PSS modified MEA could indicate neural network formation by differentiated neurons. Changes in NSC differentiation could be monitored. The method is based on non-invasive continuous measurement and so could prove a useful tool for the primary/preliminary evaluation of a pharmaceutical analysis. This article is part of a Special Issue entitled Organic Bioelectronics-Novel Applications in Biomedicine.
Product Description
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.0%
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Supelco
Potassium chloride solution, ~3 M KCl, saturated with silver chloride
Sigma-Aldrich
Potassium chloride solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%