Merck
  • Adsorption of lignocelluloses of pre-hydrolysis liquor on calcium carbonate to induce functional filler.
Adsorption of lignocelluloses of pre-hydrolysis liquor on calcium carbonate to induce functional filler.
Carbohydrate polymers (2013-04-03)
Pedram Fatehi, Fadia C Hamdan, Yonghao Ni
ABSTRACT
In this work, we aimed at adsorbing the oligo-sugars of prehydrolysis liquor on precipitated calcium carbonate (PCC) to produce modified PCC. The results showed that the adsorptions of oligo-sugars, lignin and furfural were greater on porous PCC (PCC2) than on nano-sized PCC (PCC1) due to the larger surface area of PCC2. The adsorption reached its maximum in 5 h on PCC1, but it gradually increased on PCC2 due to the diffusion of oligo-sugars and lignin into the pores of PCC2. Also, the experimental isotherm and kinetic results were well fitted into Langmuir and pseudo-second order models, respectively. The adsorption was greater at a lower temperature (i.e. 40°C) and pH (i.e. 7). Alternatively, cationic poly acrylamide (CPAM) was added to the PHL/PCC system, which led to more promising results (than that to PHL/PCC system) with the maximum lignocelluloses adsorption of 0.36 g/g on PCC2, among which 0.22 g/g was oligo-sugars.
Sigma-Aldrich
Polyacrylamide, nonionic water-soluble polymer
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Polyacrylamide, average Mn 150,000
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis