Skip to Content
Merck
All Photos(4)

Key Documents

398101

Sigma-Aldrich

Calcium carbonate

ACS reagent, 99.95-100.05% dry basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder
solubility:
dilute HCl: 6.7%
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

99.95-100.05% dry basis

form

powder

impurities

≤0.01% NH4OH pptd.
≤0.01% insol. dil. HCl

pH

8

solubility

dilute HCl: 6.7%

density

2.93 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
fluoride (F-): ≤0.0015%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.01%
Fe: ≤0.002%
K: ≤0.01%
Mg: ≤0.01%
NH4+: ≤0.003%
Na: ≤0.01%
Sr: ≤0.1%
heavy metals: ≤0.001% (by ICP)

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium carbonate is a calcium salt commonly found in shells and skeleton of marine animals.[1] It is an inexpensive, non-toxic and easily available reagent with a wide range of applications.[2]

Application

Calcium carbonate may be used:
  • As a pore generating agent to prepare porous ceramics.[3]
  • As chelometric standard for the standardization of EDTA solution.[4]
  • To extract aluminium by microwave induced thermal activation of coal fly ash (CFA).[5]

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aluminum release from microwave-assisted reaction of coal fly ash with calcium carbonate.
Zhang ZY, et al.
Fuel Processing Technology, 134, 303-309 (2015)
Opposite particle size effect on amorphous calcium carbonate crystallization in water and during heating in air.
Zou Z, et al.
Chemistry of Materials, 27(12), 4237-4246 (2015)
Kanwaljit Kaur
Handbook of Water and Wastewater Analysis, 247-248 (2007)
Calcium carbonate nanoparticles as cancer drug delivery system.
Maleki Dizaj S, et al.
Expert Opinion on Drug Delivery, 12(10) (2015)
Permeability of porous ceramic based on calcium carbonate as pore generating agent.
Sim?o L, et al.
Ceramics International, 41(3), 4782-4788 (2015)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service