Merck
  • Effect of pH, salt and chemical rinses on bacterial attachment to extracellular matrix proteins.
Effect of pH, salt and chemical rinses on bacterial attachment to extracellular matrix proteins.
Food microbiology (2013-04-02)
Siti Shahara Zulfakar, Jason D White, Tom Ross, Mark Tamplin
ABSTRACT
Microbial contamination of carcass surfaces occurs during slaughter and post-slaughter processing steps, therefore interventions are needed to enhance meat safety and quality. Although many studies have been done at the macro-level, little is known about specific processes that influence bacterial attachment to carcass surfaces, particularly the role of extracellular matrix (ECM) proteins. In the present study, the effect of pH and salt (NaCl, KCl and CaCl2) on attachment of Escherichia coli and Salmonella isolates to dominant ECM proteins: collagen I, fibronectin, collagen IV and laminin were assessed. Also, the effects of three chemical rinses commonly used in abattoirs (2% acetic acid, 2% lactic acid and 10% trisodium phosphate (TSP)) were tested. Within a pH range of 5-9, there was no significant effect on attachment to ECM proteins, whereas the effect of salt type and concentration varied depending on combination of strain and ECM protein. A concentration-dependant effect was observed with NaCl and KCl (0.1-0.85%) on attachment of E. coli M23Sr, but only to collagen I. One-tenth percent CaCl2 produced the highest level of attachment to ECM proteins for E. coli M23Sr and EC614. In contrast, higher concentrations of CaCl2 increased attachment of E. coli EC473 to collagen IV. Rinses containing TSP produced >95% reduction in attachment to all ECM proteins. These observations will assist in the design of targeted interventions to prevent or disrupt contamination of meat surfaces, thus improving meat safety and quality.
Product Description
Supelco
Potassium chloride solution, ~3 M KCl, saturated with silver chloride
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride dihydrate, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Pricing and availability is not currently available.
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, BioUltra, ≥99.5% (AT)
Pricing and availability is not currently available.
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride solution, BioUltra, ~1 M in H2O
Pricing and availability is not currently available.
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Pricing and availability is not currently available.
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Pricing and availability is not currently available.
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, ≥99.5% (KT)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Pricing and availability is not currently available.
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Pricing and availability is not currently available.