Merck
  • C-C cross-coupling of primary and secondary benzylic alcohols using supported gold-based bimetallic catalysts.
C-C cross-coupling of primary and secondary benzylic alcohols using supported gold-based bimetallic catalysts.
ChemSusChem (2013-03-28)
Xiang Liu, Ran-Sheng Ding, Lin He, Yong-Mei Liu, Yong Cao, He-Yong He, Kang-Nian Fan
ABSTRACT
Clean alcohol-alcohol cross-coupling: A clean and efficient one-pot direct C-C cross-coupling of equimolar amounts of primary and secondary alcohols by a facile hydrogen autotransfer pathway is achieved over a robust and easily recovered hydrotalcite-supported Au-Pd bimetallic catalyst system. A variety of primary and secondary alcohols have been selectively converted into the corresponding β-alkylated ketones in good yields.
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis
Sigma-Aldrich
Magnesium hydroxide, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Magnesium hydroxide, reagent grade, 95%
Sigma-Aldrich
Aluminum hydroxide, reagent grade