Merck
  • Signal amplification for electrochemical immunosensing by in situ assembly of host-guest linked gold nanorod superstructure on immunocomplex.
Signal amplification for electrochemical immunosensing by in situ assembly of host-guest linked gold nanorod superstructure on immunocomplex.
Biosensors & bioelectronics (2013-03-19)
Dajie Lin, Jie Wu, Huangxian Ju, Feng Yan
ABSTRACT
An amplification strategy for signal tracing was developed by introducing a host-guest binding reaction into the assembly process of gold nanorods (AuNRs) superstructure. The amplification pathway firstly used a thio-β-cyclodextrin (SH-β-CD) functionalized gold nanoparticles to label signal antibody, and then in situ assembled multi-layer SH-β-CD end-functionalized AuNRs to sandwich immunocomplex on immunosensor surface by using 4,4,4,4-(21H, 23H-porphine-5,10,15,20-tetrayl) tetrakis (benzoic acid) as a bridge to achieve simple and convenient host-guest reaction. The built end-to-end AuNRs superstructure showed excellent performance for the signal amplification in connection with the electrochemical biosensing by preoxidation and then voltammetric analysis of gold element. Using α-fetoprotein as an analyte, the immunosensor was constructed by covalently binding capture antibody to chitosan-carbon nanotubes-poly(diallyldimethylammonium chloride) modified electrode. The superstructure rich in AuNRs brought an enhanced detection sensitivity of protein, which could detect α-fetoprotein in a linear range from 0.5 pg mL(-1) to 0.5 ng mL(-1) with a detection limit down to 0.032 pg mL(-1). The immunoassay exhibited good stability and acceptable reproducibility and accuracy. The in situ superstructure assembly could be extended to other labeled recognition systems, providing a promising novel avenue for signal amplification and potential applications in bioanalysis and clinical diagnostics.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
Potassium benzoate, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%