Skip to Content
Merck
All Photos(1)

Key Documents

73983

Supelco

Mettler-Toledo Calibration substance ME 18555, Benzoic acid

Mettler-Toledo Calibration substance ME 18555, Benzoic acid

analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)

Synonym(s):

Benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$234.00

$234.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$234.00

About This Item

Linear Formula:
C6H5COOH
CAS Number:
Molecular Weight:
122.12
Beilstein:
636131
EC Number:
MDL number:
UNSPSC Code:
20121904
PubChem Substance ID:
NACRES:
NA.24

$234.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

4.21 (vs air)

vapor pressure

10 mmHg ( 132 °C)

autoignition temp.

1061 °F

quality

traceable to primary standards (LGC)

shelf life

limited shelf life, expiry date on the certificate and label

bp

249 °C (lit.)

mp

121-125 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

SMILES string

OC(=O)c1ccccc1

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)

InChI key

WPYMKLBDIGXBTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mettler-Toledo calibration standard ME 18555, benzoic acid (121-125 °C) is suitable for the regular calibration of Mettler-Toledo melting point instrument. Its melting point value is an average of 6 to 12 measurements with a Mettler-Toledo MP90 excellence melting point instrument that is calibrated against primary standards.

Application

This Mettler-Toledo melting point standard is used for the calibration of the melting point instrument Mettler FP900 thermosystem.

Features and Benefits

  • Melting point calibration standard traceable to a primary standard from LGC, London
  • Melting point evaluation performed in both thermodynamic and pharmacopeia modes
  • Available with certificates of analysis and safety data sheet
  • A product of analytical standard grade to help meet the QC/QA requirements of melting point analysis

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davidson M.P, et al.
Antimicrobials in Food, (3) (2005)
Final report on the safety assessment of Benzyl Alcohol, Benzoic Acid, and Sodium Benzoate
NB
International Journal of Toxicology, 20, 23-50 (2001)
S Nacht et al.
Journal of the American Academy of Dermatology, 4(1), 31-37 (1981-01-01)
The transepidermal penetration and metabolic disposition of 14C-benzoyl peroxide were assessed in vitro (excised human skin) and in vivo (rhesus monkey). In vitro, the benzoyl peroxide penetrated into the skin, through the stratum corneum or the follicular openings, or both
Copper-mediated C-H/C-H biaryl coupling of benzoic acid derivatives and 1,3-azoles.
Mayuko Nishino et al.
Angewandte Chemie (International ed. in English), 52(16), 4457-4461 (2013-03-21)
Ly Dieu Tran et al.
Journal of the American Chemical Society, 134(44), 18237-18240 (2012-10-30)
An auxiliary-assisted, copper catalyzed or promoted sulfenylation of benzoic acid derivative β-C-H bonds and benzylamine derivative γ-C-H bonds has been developed. The method employs disulfide reagents, copper(II) acetate, and DMSO solvent at 90-130 °C. Application of this methodology to the

Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. Are the Mettler-Toledo Calibration melting standards already dried and pulverized, ready to use out of the bottle?

    1 answer
    1. The answer is yes. All melting point standards are ground to a fine powder, tested as melting point standards and ready to use.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service