Merck
  • Enhanced mechanical properties and blood compatibility of PDMS/liquid crystal cross-linked membrane materials.
Enhanced mechanical properties and blood compatibility of PDMS/liquid crystal cross-linked membrane materials.
Journal of the mechanical behavior of biomedical materials (2013-03-05)
Huaxin Rao, Ziyong Zhang, Fanna Liu
ABSTRACT
A novel polydimethylsiloxane/liquid crystal cross-linked membrane (PDMS/LC) was prepared by using PDMS containing vinyl groups and LCs containing unsaturated linkages as matrix materials. Mechanical properties, liquid crystalline performance and blood compatibility of the PDMS/LC cross-linked membrane containing different LC contents and LC groups were investigated, respectively. The results showed that mechanical properties of the membrane increased more significantly than those of pure PDMS membranes. The PDMS/LC cross-linked membrane also possessed better membrane-forming ability, lower hemolysis rate, less platelets adhesion and more favorable anti-coagulant properties. Additionally, mechanical properties and blood compatibility of the membrane can be enhanced simultaneously and obviously due to the introduction of the cholesteric liquid crystals and the application of the preferred cross-linked reaction without byproducts.
Sigma-Aldrich
Silicone oil, viscosity 5 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 500 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 20 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 50 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 30,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil
Silicone oil, for melting point and boiling point apparatuses
Sigma-Aldrich
Silicone oil, viscosity 100,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 60,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 1,000 cSt (25 °C)