Merck
  • Natural product derivatives with bactericidal activity against Gram-positive pathogens including methicillin-resistant Staphylococcus aureus and vancomycin-resistant Enterococcus faecalis.
Natural product derivatives with bactericidal activity against Gram-positive pathogens including methicillin-resistant Staphylococcus aureus and vancomycin-resistant Enterococcus faecalis.
Bioorganic & medicinal chemistry letters (2010-08-31)
Joshua B Phillips, Adrienne E Smith, Brian R Kusche, Bradley A Bessette, P Whitney Swain, Stephen C Bergmeier, Mark C McMills, Dennis L Wright, Nigel D Priestley
ABSTRACT
We have shown that the intentional engineering of a natural product biosynthesis pathway is a useful way to generate stereochemically complex scaffolds for use in the generation of combinatorial libraries that capture the structural features of both natural products and synthetic compounds. Analysis of a prototype library based upon nonactic acid lead to the discovery of triazole-containing nonactic acid analogs, a new structural class of antibiotic that exhibits bactericidal activity against drug resistant, Gram-positive pathogens including Staphylococcus aureus and Enterococcus faecalis.
Sigma-Aldrich
Nonactin, from Streptomyces griseus, ≥98.0% (Total homologs, HPLC)
Sigma-Aldrich
Ammonium ionophore I, Selectophore, function tested, ≥98.0% ((Total homologs), HPLC)