Merck
  • Development and validation of stability-indicating TLC-densitometric method for determination of betaxolol with LC-ESI/MS analysis of degradation product.
Development and validation of stability-indicating TLC-densitometric method for determination of betaxolol with LC-ESI/MS analysis of degradation product.
Acta poloniae pharmaceutica (2013-08-08)
Anna Kwiecień, Jan Krzek, Maria Walczak, Mateusz Mazur
ABSTRACT
The purpose of this work was to develop a sensitive stability indicating TLC-densitometric method for the determination of betaxolol (Bx) in pharmaceutical preparations and to study the stability of Bx in acidic solutions. The method was developed on TLC aluminium plates precoated with silica gel F254using the mobile phase chloroform-methanol-ammonia 25% (18 : 4: 0.2, v/v/v) which gives compact spots for Bx (R(f) approximately equal to 0.64) and its degradation product (R(f) approximately equal to 0.39). Densitometric analysis was carried out in UV at 280 nm. The developed method is highly sensitive (LOD = 66.6 ng/spot, LOQ = 200 ng/spot), precise (RSD = 2.73%) and accurate (mean recovery = 100.28% at 100% level). Bx was subjected to acidic and alkaline hydrolysis but degradation was observed only in acidic solutions. The degradation process was described with kinetic and thermodynamic parameters. Based on LC-ESI/MS analysis, it was found that Bx decomposes in acidic solution to produce ethoxyphenoxy-3-[(1-methylethyl)amino]propan-2-ol.
Product Description
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane