Merck
  • [Approximately four years of survival benefits for a case of gastric cancer postoperative carcinomatous peritonitis by combined modality therapy].
[Approximately four years of survival benefits for a case of gastric cancer postoperative carcinomatous peritonitis by combined modality therapy].
Gan to kagaku ryoho. Cancer & chemotherapy (2013-07-19)
Kazuhisa Katayama, Takashi Nishino, Yuji Takahashi, Takashi Tanaka, Hiroshi Okitsu, Natsu Okitsu
ABSTRACT
Carcinomatous peritonitis may develop after operation for gastric cancer. As ascites are difficult to control, especially for gastric cancer postoperative carcinomatous peritonitis, many cases are difficult to treat. The present case was a female patient with carcinomatous peritonitis that occurred 2 years post-surgery. Administration of docetaxel (DOC)and S-1 combination therapy achieved a complete response. However, she had a relapse of carcinomatous peritonitis 3 years post-surgery. She underwent bypass operation, followed by DOC and S-1 combination therapy again. She achieved a good quality of life for more than two years. As side effects in patients worsen with the repeated exposure to chemotherapy, continuing the same treatment is difficult. Therefore, we changed the therapy method to irinotecan(CPT-11)/cisplatin(CDDP)therapy, weekly paclitaxel(PTX)and methotrexate(MTX)/5-fluorouracil(5-FU)therapy, and bypass operation when necessary. Rapid progression of her condition was sequentially suppressed, allowing her to continue her everyday life. Overall, this treatment method provided survival benefits of approximately four years following the recurrence of carcinomatous peritonitis.
Sigma-Aldrich
Tegafur, ≥98% (HPLC), powder
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Irinotecan hydrochloride, topoisomerase inhibitor
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
7-Ethyl-10-hydroxycamptothecin, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Paclitaxel semi-synthetic for system suitability, European Pharmacopoeia (EP) Reference Standard
Paclitaxel, European Pharmacopoeia (EP) Reference Standard
Methotrexate, European Pharmacopoeia (EP) Reference Standard