Merck
Mifepristone as a therapeutic agent in psychiatry.
Journal of psychosocial nursing and mental health services (2013-07-03)
Robert H Howland
ABSTRACT
The hypothalamic-pituitary-adrenal (HPA) axis is the body's main stress-response system, and cortisol is the major adrenal glucocorticoid hormone secreted in human beings. HPA axis activity and cortisol secretion is regulated by a negative feedback system involving glucocorticoid receptors. Dysregulation of the HPA axis and increased cortisol levels have been implicated in mood, psychotic, and other psychiatric disorders. Mifepristone, as a potent antagonist of glucocorticoid receptors, has been studied or is currently being investigated as a potential therapeutic agent for psychotic depression, posttraumatic stress disorder, and alcohol and cocaine dependence, as well as for mitigating the weight gain associated with the use of antipsychotic drugs and for improving cognitive dysfunction in schizophrenia and bipolar disorder. This article will review some of the work in these areas.
Sigma-Aldrich
Mifepristone, ≥98%
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material