Merck
  • [Preliminary study on cultivation of adventitious roots of Hypericum perforatum in bioreactors].
[Preliminary study on cultivation of adventitious roots of Hypericum perforatum in bioreactors].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-05-01)
Xiao-Kun Yu, Xuan-Chun Piao, Yue Dai, Tie-Jun Li, Mei-Lan Lian
ABSTRACT
To cultivate adventitious roots of Hypericum perforatum in bioreactors, in order to seek for suitable conditions for adventitious growth. The effect of IBA concentration, sugar type and concentration, inoculum volume and air volume of adventitious roots on the cultivation of adventitious roots of H. perforatum was observed in a 5 L air-lift bioreactor. Adventitious roots of H. perforatum were cultivated in a MS culture dish. With the increase of IBA concentration, the propagation coefficient of adventitious roots of H. perforatum was on the rise. The IBA concentration ranging between 1.25-1.75 mg x L(-1) was suitable for the growth of adventitious roots. Adventitious roots grew best with sucrose in MS medium, with the propagation coefficient up to 22.15. When sucrose concentration was 30 g x L(-1), fresh weight, dry weight and propagation coefficient reached the maximum value. An adventitious root reactor with an inoculum volume of 20 g was favorable for the growth of adventitious roots. The air volume of reactors of 0.075 vvm (air volume/culture volume per minute) was favorable for the growth of adventitious roots, with the significant increase in the propagation coefficient of adventitious roots. In the amplification experiment, we found that the cultivation conditions of adventitious roots in a 5 L bioreactor was completely applicable to that in 10 and 20 L bioreactors, and adventitious roots grew well in a large bioreactor. IBA concentration, sugar type and concentration, inoculum volume and air volume had a significant effect on the growth of adventitious roots.
Product Description
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Indole-3-butyric acid, ≥98.0% (T)
Sigma-Aldrich
Indole-3-butyric acid, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Indole-3-butyric acid potassium salt, suitable for plant cell culture, BioReagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)