Merck
  • Electrocatalytic degradation kinetic of 4-chlorophenol by the Pd/C gas-diffusion electrode system.
Electrocatalytic degradation kinetic of 4-chlorophenol by the Pd/C gas-diffusion electrode system.
Water science and technology : a journal of the International Association on Water Pollution Research (2013-04-13)
Z Y Bian, Y Bian, H Wang, L Pang, A Z Ding
ABSTRACT
A Pd/C gas-diffusion cathode which generated H2O2 through a two-electron reduction process of fed oxygen molecule was used to degrade 4-chlorophenol in an undivided electrolysis device. The kinetics of 4-chlorophenol degradation has been investigated by the electrochemical oxidation processes. By inspecting the relationship between the rate constants (k) and influencing factors, using first-order kinetics to describe the electrochemical oxidation process of 4-chlorophenol, a kinetic model of 4-chlorophenol degradation process was proposed to calculate the 4-chlorophenol effluent concentration: C = C0 exp( -3:76 × 10(-6) C(-0.5)0 J(2) M(-0.7) Q(0.17) Dt). It was found that the electrocatalytic degradation rate of 4-chlorophenol was affected by current density, electrode distance, air-feeding rate, electrolyte concentration and initial 4-chlorophenol concentration. The kinetics obtained from the experiments under corresponding electrochemical conditions could provide an accurate estimation of 4-chlorophenol effluent concentration and lead to better design of the electrochemical reactor.
Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
4-Chlorophenol, ≥99%