Merck
  • DNA methylation potential: dietary intake and blood concentrations of one-carbon metabolites and cofactors in rural African women.
DNA methylation potential: dietary intake and blood concentrations of one-carbon metabolites and cofactors in rural African women.
The American journal of clinical nutrition (2013-04-12)
Paula Dominguez-Salas, Sophie E Moore, Darren Cole, Kerry-Ann da Costa, Sharon E Cox, Roger A Dyer, Anthony J C Fulford, Sheila M Innis, Robert A Waterland, Steven H Zeisel, Andrew M Prentice, Branwen J Hennig
ABSTRACT
Animal models show that periconceptional supplementation with folic acid, vitamin B-12, choline, and betaine can induce differences in offspring phenotype mediated by epigenetic changes in DNA. In humans, altered DNA methylation patterns have been observed in offspring whose mothers were exposed to famine or who conceived in the Gambian rainy season. The objective was to understand the seasonality of DNA methylation patterns in rural Gambian women. We studied natural variations in dietary intake of nutrients involved in methyl-donor pathways and their effect on the respective metabolic biomarkers. In 30 women of reproductive age (18-45 y), we monitored diets monthly for 1 y by using 48-h weighed records to measure intakes of choline, betaine, folate, methionine, riboflavin, and vitamins B-6 and B-12. Blood biomarkers of these nutrients, S-adenosylhomocysteine (SAH), S-adenosylmethionine (SAM), homocysteine, cysteine, and dimethylglycine were also assessed monthly. Dietary intakes of riboflavin, folate, choline, and betaine varied significantly by season; the most dramatic variation was seen for betaine. All metabolic biomarkers showed significant seasonality, and vitamin B-6 and folate had the highest fluctuations. Correlations between dietary intakes and blood biomarkers were found for riboflavin, vitamin B-6, active vitamin B-12 (holotranscobalamin), and betaine. We observed a seasonal switch between the betaine and folate pathways and a probable limiting role of riboflavin in these processes and a higher SAM/SAH ratio during the rainy season. Naturally occurring seasonal variations in food-consumption patterns have a profound effect on methyl-donor biomarker status. The direction of these changes was consistent with previously reported differences in methylation of metastable epialleles. This trial was registered at www.clinicaltrials.gov as NCT01811641.
Sigma-Aldrich
Sarcosine, 98%
Sigma-Aldrich
S-(5′-Adenosyl)-L-homocysteine, crystalline
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Sarcosine, BioXtra
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
N,N-Dimethylglycine hydrochloride
Sigma-Aldrich
Sarcosine, crystallized, ≥98.0% (T)
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Homocysteine, ≥98.0% (NT)
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Supelco
Cyanocobalamin (B12), analytical standard
Supelco
Riboflavin (B2), analytical standard
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Sigma-Aldrich
DL-Methionine, ≥99.0% (NT)
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
N,N-Dimethylglycine, ≥99%
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
Betaine hydrochloride, ≥99%