Merck
  • At very low concentrations known chaotropes act as kosmotropes for the N and B isoforms of human serum albumin.
At very low concentrations known chaotropes act as kosmotropes for the N and B isoforms of human serum albumin.
Biochemistry and cell biology = Biochimie et biologie cellulaire (2013-03-27)
Priyankar Sen, Mohd Moin Khan, Asif Equbal, Ejaz Ahmad, Rizwan Hasan Khan
ABSTRACT
Very few studies have been done to understand the effect of millimolar concentrations of chaotropes on protein structure. In our previous study we observed that the secondary and tertiary structure of human serum albumin (HSA) increases in the presence of 5 mmol/L urea. Micelle formation in amphoteric detergents increases in the presence of equivalent concentrations of urea. Here, we observed a significant increase in the secondary and tertiary structure of HSA. Interestingly, guanidine hydrochloride, another chaotropic agent, also shows a similar effect. Our results show electrostatic interaction may play a role in neutral to basic transition in HSA. This study further supports the claim that at millimolar concentrations the chaotropes may act as kosmotropes for proteins.
Sigma-Aldrich
Guanidine sulfate salt, 99%
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)