Merck
  • Cloud point extraction and flame atomic absorption spectrometry determination of lead (II) in environmental and food samples.
Cloud point extraction and flame atomic absorption spectrometry determination of lead (II) in environmental and food samples.
Journal of AOAC International (2013-03-05)
Mustafa Soylak, Erkan Yilmaz, Mehrorang Ghaedi, Mortaza Montazerozohori, Marjan Sheibani
ABSTRACT
A cloud point extraction procedure for the preconcentration of Pb2+ in various samples following complexation with 2,2'-(1E,1'E)-1,1'-(2,2'-azanediylbis(ethane-2,1-diyl)bis(azan-1-yl-1-ylidene)) bis(ethan-1-yl-1-ylidene)diphenol in Triton X-114 after centrifugation is reported. A 0.5 mL portion of methanol acidified with 1.0 M HNO3 was added to the surfactant-rich phase prior to analysis by flame atomic absorption spectrometry. The influence of analytical parameters--including pH, concentrations of ligand, Triton X-114, and HNO3, bath temperature, heating time, and centrifugation rate and time--were optimized, and the effect of the matrix ions on the recovery of Pb2+ was investigated. An LOD of 1.9 ng/mL along with a preconcentration factor of 50 with RSD of 1.0% for Pb2+ were achieved. The proposed procedure was applied to the analysis of various real samples.
Product Description
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Triton X-405 solution, 70% in H2O
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥65%
Sigma-Aldrich
Triton X-114
Supelco
IGEPAL® CA-630
Sigma-Aldrich
IGEPAL® CA-720, average Mn ~735
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Sigma-Aldrich
Triton X-114, laboratory grade
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
Triton X-165 solution, Non-ionic, CMC-0.43 mM, suitable for electrophoresis, cloud point >100 °C