Merck
  • Nickel-catalyzed cross-couplings of benzylic pivalates with arylboroxines: stereospecific formation of diarylalkanes and triarylmethanes.
Nickel-catalyzed cross-couplings of benzylic pivalates with arylboroxines: stereospecific formation of diarylalkanes and triarylmethanes.
Journal of the American Chemical Society (2013-02-22)
Qi Zhou, Harathi D Srinivas, Srimoyee Dasgupta, Mary P Watson
ABSTRACT
We have developed a stereospecific nickel-catalyzed cross-coupling of benzylic pivalates with arylboroxines. The success of this reaction relies on the use of Ni(cod)2 as the catalyst and NaOMe as a uniquely effective base. This reaction has broad scope with respect to the arylboroxine and benzylic pivalate, enabling the synthesis of a variety of diarylalkanes and triarylmethanes in good to excellent yields and ee's.
Sigma-Aldrich
Pivalic acid, 99%