Merck
  • Consequences of normalizing transcriptomic and genomic libraries of plant genomes using a duplex-specific nuclease and tetramethylammonium chloride.
Consequences of normalizing transcriptomic and genomic libraries of plant genomes using a duplex-specific nuclease and tetramethylammonium chloride.
PloS one (2013-02-15)
Marta Matvienko, Alexander Kozik, Lutz Froenicke, Dean Lavelle, Belinda Martineau, Bertrand Perroud, Richard Michelmore
ABSTRACT
Several applications of high throughput genome and transcriptome sequencing would benefit from a reduction of the high-copy-number sequences in the libraries being sequenced and analyzed, particularly when applied to species with large genomes. We adapted and analyzed the consequences of a method that utilizes a thermostable duplex-specific nuclease for reducing the high-copy components in transcriptomic and genomic libraries prior to sequencing. This reduces the time, cost, and computational effort of obtaining informative transcriptomic and genomic sequence data for both fully sequenced and non-sequenced genomes. It also reduces contamination from organellar DNA in preparations of nuclear DNA. Hybridization in the presence of 3 M tetramethylammonium chloride (TMAC), which equalizes the rates of hybridization of GC and AT nucleotide pairs, reduced the bias against sequences with high GC content. Consequences of this method on the reduction of high-copy and enrichment of low-copy sequences are reported for Arabidopsis and lettuce.
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥97%
Supelco
Tetramethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium iodide, 99%
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
Sigma-Aldrich
Tetramethylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Tetramethylammonium chloride solution, for molecular biology
Sigma-Aldrich
Tetramethylammonium bromide, 98%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Sigma-Aldrich
Tetramethylammonium chloride, reagent grade, ≥98%
Supelco
Tetramethylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium fluoride, 97%
Sigma-Aldrich
Tetramethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Tetramethylammonium sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent