Merck
  • Synthesis and characterization of an azo dibenzoic acid Schiff base and its Ni(II), Pb(II), Zn(II) and Cd(II) complexes.
Synthesis and characterization of an azo dibenzoic acid Schiff base and its Ni(II), Pb(II), Zn(II) and Cd(II) complexes.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-02-05)
Ali Kakanejadifard, Fatemeh Esna-ashari, Payman Hashemi, Abedin Zabardasti
ABSTRACT
The new Schiff base 4,4'-(1E,1'E)-(3,3'-(1E,1'E)-(pyridine-2,6-diylbis(azan-1-yl-1-ylid ene))bis(methan-1-yl-1-ylidene)bis(4-hydroxy-3,1-phenylene))bis(diazene-2,1-diyl)dibenzoic acid (1) was prepared from the condensation reaction of 2,6-diaminopyridine with 4-((3-formyl-4-hydroxyphenyl)diazenyl)benzoic acid in methanol. The compound 1 is potentially an N, O multidentate chelating ligand which could form stable complexes with metal ions in 1:1 up to 1:3mol ratio of metal to ligand. The 1:1 complexes of Schiff base 1 with Ni(II), Pb(II), Zn(II) and Cd(II) have been synthesized by its condensation reaction with appropriate salts of metal ions. Structures of Schiff base (1) as well as its complexes with abovementioned metal ions were characterized by elemental analysis, mass, IR, UV-vis., (1)H and (13)С NMR spectroscopy.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Potassium benzoate, ReagentPlus®, 99%
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland