Merck
  • A new bis-tetraamine ligand with a chromophoric 4-(9-anthracenyl)-2,6-dimethylpyridinyl linker for glyphosate and ATP sensing.
A new bis-tetraamine ligand with a chromophoric 4-(9-anthracenyl)-2,6-dimethylpyridinyl linker for glyphosate and ATP sensing.
Dalton transactions (Cambridge, England : 2003) (2013-02-02)
Jacky Pouessel, Sabah Abada, Nathalie Le Bris, Mourad Elhabiri, Loïc J Charbonnière, Raphaël Tripier
ABSTRACT
The synthesis of a new linear bis-tetraamine ligand L1, based on two 1,4,8,11-tetraazaundecane units grafted at the 2 and 6 positions of a pyridinyl linker substituted by an anthracenyl fluorophore in the para position, is described and anion complexation studies of L1 with anionic substrates are reported. The protonation pattern and the study of the binding properties of L1 in an aqueous medium with two anionic substrates, the nucleotide adenosine triphosphate (ATP) and the herbicide glyphosate (N-(phosphonomethyl)glycine, PMG), were investigated by means of potentiometry, NMR spectroscopy and absorption and emission spectroscopic techniques. To decipher the impact of the chromophoric linker on the complexation process and to highlight its optical properties, a comparison is established with its previously reported analog L2 devoid of the anthracenyl group. The results unambiguously show that the protonation and complexation properties are preserved despite the presence of the bulky linker, allowing for the use of L1 as a fluorescent sensor for ATP and PMG.
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Supelco
Anthracene, analytical standard
Supelco
Pyridine solution, certified reference material, 2000 μg/mL in methanol
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Pyridine hydrochloride, purum, ≥98.0% (AT)
Sigma-Aldrich
Pyridine hydrochloride, 98%
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Sigma-Aldrich
Anthracene, reagent grade, 97%
Sigma-Aldrich
N-(Phosphonomethyl)glycine, BioReagent, suitable for plant cell culture
Sigma-Aldrich
N-(Phosphonomethyl)glycine, 96%
Supelco
Anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Pyridine, analytical standard
Supelco
Glyphosate, PESTANAL®, analytical standard
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine hydrobromide, 98%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%