Merck
  • Interferences in the direct quantification of bisphenol S in paper by means of thermochemolysis.
Interferences in the direct quantification of bisphenol S in paper by means of thermochemolysis.
Journal of chromatography. A (2013-01-12)
Valentina Becerra, Jürgen Odermatt
ABSTRACT
This article analyses the interferences in the quantification of traces of bisphenol S in paper by applying the direct analytical method "analytical pyrolysis gas chromatography mass spectrometry" (Py-GC/MS) in conjunction with on-line derivatisation with tetramethylammonium hydroxide (TMAH). As the analytes are simultaneously analysed with the matrix, the interferences derive from the matrix. The investigated interferences are found in the analysis of paper samples, which include bisphenol S derivative compounds. As the free bisphenol S is the hydrolysis product of the bisphenol S derivative compounds, the detected amount of bisphenol S in the sample may be overestimated. It is found that the formation of free bisphenol S from the bisphenol S derivative compounds is enhanced in the presence of tetramethylammonium hydroxide (TMAH) under pyrolytic conditions. In order to avoid the formation of bisphenol S trimethylsulphonium hydroxide (TMSH) is introduced. Different parameters are optimised in the development of the quantification method with TMSH. The quantification method based on TMSH thermochemolysis has been validated in terms of reproducibility and accuracy.
Product Description
Supelco
Tetramethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Trimethylsulfonium iodide, 98%
Sigma-Aldrich
Tetramethylammonium iodide, 99%
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥97%
Sigma-Aldrich
Tetramethylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Tetramethylammonium chloride solution, for molecular biology
Sigma-Aldrich
Tetramethylammonium bromide, 98%
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Sigma-Aldrich
Tetramethylammonium chloride, reagent grade, ≥98%
Supelco
Tetramethylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium fluoride, 97%
Sigma-Aldrich
Tetramethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Tetramethylammonium sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Sigma-Aldrich
4,4′-Sulfonyldiphenol, 98%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O