Merck
  • Enhancement of anti-cholinesterase activity of Zingiber cassumunar essential oil using a microemulsion technique.
Enhancement of anti-cholinesterase activity of Zingiber cassumunar essential oil using a microemulsion technique.
Drug discoveries & therapeutics (2012-12-12)
S Okonogi, W Chaiyana
ABSTRACT
The aim of the present study was to enhance the cholinesterase inhibitory activity of Zingiber cassumunar (ZC) oil using a microemulsion (ME) technique. Pseudoternary phase diagrams of the oil, water, and surfactant/co-surfactant mixture were constructed using a water titration method. Effects of co-surfactant, surfactant/co-surfactant ratio, ionic strength, and pH were examined by means of the microemulsion region which existed in the phase diagrams. The inhibition of acetylcholinesterase (AChE) and butyrylcholinesterase (BChE) were tested by Ellman's colorimetric assay. It was found that ZC oil possesses inhibitory activity against not only AChE but also BChE. Formulation of ZC oil as ME revealed that alkyl chain length and number of hydroxyl groups of co-surfactant exhibited a remarkable effect on the pseudoternary phase diagram. Longer alkyl chains and more hydroxyl groups gave smaller regions of MEs. Ionic strength also affected the ME region. However, the phase behavior was hardly influenced by pH. The suitable ZC oil ME was composed of Triton X-114 in combination with propylene glycol. The anti-cholinesterase activity of this ME was much higher than that of native ZC oil. It exhibited twenty times and twenty five times higher inhibitory activity against AChE and BChE, respectively. ZC oil loaded ME is an attractive formulation for further characterization and an in vivo study in an animal model with Alzheimer's disease.
Sigma-Aldrich
Triton X-114
Supelco
IGEPAL® CA-630
Sigma-Aldrich
(R)-(−)-1,2-Propanediol, 96%
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
IGEPAL® CA-720, average Mn ~735
Sigma-Aldrich
Triton X-114, laboratory grade
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
Triton X-165 solution, Non-ionic, CMC-0.43 mM, suitable for electrophoresis, cloud point >100 °C
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
(S)-(+)-1,2-Propanediol, 96%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
Triton X-405 solution, 70% in H2O
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%