Merck
  • Non-edible babassu oil as a new source for energy production-a feasibility transesterification survey assisted by ultrasound.
Non-edible babassu oil as a new source for energy production-a feasibility transesterification survey assisted by ultrasound.
Ultrasonics sonochemistry (2012-12-05)
Eduardo J M Paiva, Maria Lucia C P da Silva, Jayne C S Barboza, Pedro C de Oliveira, Heizir F de Castro, Domingos S Giordani
ABSTRACT
Ethyl esters of babassu oil were synthesized by alkaline catalysis to make the green production of biodiesel feasible with simple methods and available technology. Babassu oil is a transparent, light yellow oil extracted from the seeds of the babassu palm (Orbinya sp), and due to its high saturated fatty acid composition (83%), it is considered a non-inedible oil. Transesterification using ethanol represents a valid alternative to using methanol because of ethanol's lower toxicity and the higher yield on weight compared to methanol. Statistical methodology was applied to optimize the transesterification reaction, which was promoted by ultrasonic waves and mechanical agitation. Nuclear magnetic resonance spectroscopy was used to quantify the conversion attained. Alkaline transesterification assisted by ultrasound produced the best results with respect to reaction time and the phase separation step. The model obtained showed that conversions higher than 97% may be achieved in 10min with correct tuning of the process variables.
Product Description
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Supelco
Palm oil, analytical standard
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets