Merck
  • Versatile peroxidase of Bjerkandera fumosa: substrate and inhibitor specificity.
Versatile peroxidase of Bjerkandera fumosa: substrate and inhibitor specificity.
Enzyme and microbial technology (2012-12-04)
Natalia Pozdnyakova, Oleg Makarov, Marina Chernyshova, Olga Turkovskaya, Anna Jarosz-Wilkolazka
ABSTRACT
The inhibitor and substrate specificities of versatile peroxidase from Bjerkandera fumosa (VPBF) were studied. Two different effects were found: NaN(3), Tween-80, anthracene, and fluorene decreased the activity of VPBF, but p-aminobenzoic acid increased it. A mixed mechanism of effector influence on the activity of this enzyme was shown. The catalytic properties of VPBF in the oxidation of mono- and polycyclic aromatic compounds were studied also. 2,7-Diaminofluorene, ABTS, veratryl alcohol, and syringaldazine can be oxidized by VPBF in two ways: either directly by the enzyme or by diffusible chelated Mn(3+) as an oxidizing agent. During VPBF oxidation of 2,7-diaminofluorene, both with and without Mn(2+), biphasic kinetics with apparent saturation in both micromolar and millimolar ranges were obtained. In the case of ABTS, inhibition of VPBF activity by an excess of substrate was observed. Direct oxidation of p-aminobenzoic acid by versatile peroxidase was found for the first time. The oxidation of three- and four-ring PAHs by VPBF was investigated, and the oxidation of anthracene, phenanthrene, fluorene, pyrene, chrysene, and fluoranthene was shown. The products of PAH oxidation (9,10-anthraquinone, 9,10-phenanthrenequinone, and 9-fluorenone) catalyzed by VPBF were identified.
Sigma-Aldrich
Sodium azide 0.1 M solution
Supelco
Anthracene, analytical standard
Supelco
Anthracene, analytical standard
Supelco
Fluorene, analytical standard
Sigma-Aldrich
2,6-Dimethoxyphenol, natural, ≥96%
Supelco
4-Aminobenzoic acid, analytical standard
Supelco
Pyrogallol, analytical standard
Supelco
2,6-Dimethoxyphenol, analytical standard
Supelco
Anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
4-Aminobenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
4-Aminobenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
2,6-Dimethoxyphenol, ≥98%, FG
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Sigma-Aldrich
Anthracene, reagent grade, 97%
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Sigma-Aldrich
4-Aminobenzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Pyrogallol, ACS reagent
Sigma-Aldrich
2,6-Dimethoxyphenol, 99%
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Sigma-Aldrich
Fluorene, 98%