Merck
Stealth rare earth oxide nanodiscs for magnetic resonance imaging.
Advanced healthcare materials (2012-11-28)
Mark J Bailey, Rob van der Weegen, Piper J Klemm, Suzanne L Baker, Brett A Helms
ABSTRACT
Configuring RE(2) O(3) nanocrystals with pseudo-2D morphologies confers substantive gains in relaxivities over equivalent spherical counterparts. The most promising arises from Gd(2) O(3) whose ionic transverse and longitudinal relaxivities were as high as r(1) = 12.7 mM(-1) s(-1) and r(2) = 17.2 mM(-1) s(-1) (dispersed in H(2) O, T = 37 °C & B(0) = 1.41 T), pointing to new opportunities to achieve high contrast MRI while concomitantly affording longer half-life potential in vivo.
Sigma-Aldrich
Sodium perrhenate, 99.99% trace metals basis
Sigma-Aldrich
Ammonium perrhenate, ≥99%
Sigma-Aldrich
Ammonium perrhenate, 99.999% trace metals basis
Sigma-Aldrich
Potassium perrhenate, 99.98% trace metals basis
Sigma-Aldrich
Potassium perrhenate, 99%