Merck
  • A facile one-pot solvothermal method to produce superparamagnetic graphene-Fe3O4 nanocomposite and its application in the removal of dye from aqueous solution.
A facile one-pot solvothermal method to produce superparamagnetic graphene-Fe3O4 nanocomposite and its application in the removal of dye from aqueous solution.
Colloids and surfaces. B, Biointerfaces (2012-09-27)
Qiuhua Wu, Cheng Feng, Chun Wang, Zhi Wang
ABSTRACT
A superparamagnetic graphene-Fe(3)O(4) nanocomposite (G/Fe(3)O(4)) was synthesized by a facile one-pot solvothermal method. The nanocomposite G/Fe(3)O(4) prepared by the new method was firstly used as an adsorbent to remove dye for water pollution remediation. In comparison with G/Fe(3)O(4) prepared by the in situ chemical coprecipitation, the newly prepared G/Fe(3)O(4) had a higher adsorption efficiency for the dye. The adsorption characteristics of the nanocomposite adsorbent were examined using the organic dye pararosaniline as the adsorbate. The adsorption kinetics, adsorption capacity of the adsorbent, and the effect of the adsorbent dosage and solution pH on the removal efficiency of pararosaniline were investigated. The adsorption capacity of G/Fe(3)O(4) for pararosaniline was evaluated using the Freundlich and Langmuir adsorption isotherm models. The G/Fe(3)O(4) hybrid composite can be easily manipulated in magnetic field for desired separation, leading to an easy removal of the dye from polluted water. The G/Fe(3)O(4) hybrid composite would have a great potential in removing organic dyes from polluted water.
Sigma-Aldrich
Pararosaniline hydrochloride, >85.0% (HPLC)
Sigma-Aldrich
Basic Fuchsin, certified by the BSC, Dye content ≥88 %
Sigma-Aldrich
Basic Fuchsin, Dye content >85 %
Sigma-Aldrich
Pararosaniline acetate, Dye content 90 %