Merck
  • A nonradioisotope chemiluminescent assay for evaluation of 2-deoxyglucose uptake in 3T3-L1 adipocytes. Effect of various carbonyls species on insulin action.
A nonradioisotope chemiluminescent assay for evaluation of 2-deoxyglucose uptake in 3T3-L1 adipocytes. Effect of various carbonyls species on insulin action.
Biochimie (2012-07-28)
Nicolas Vidal, Jean Pierre Cavaillé, Marjorie Poggi, Franck Peiretti, Pierre Stocker
ABSTRACT
We have developed a rapid nonradioisotope chemiluminescent assay adapted to high-throughput screening experiments, to evaluate glucose uptake activity in cultured cells. For chemiluminescence quantification of 2-deoxyglucose, we used a luminol oxidation reaction after an enzymatic dephosphorylation of 2-deoxyglucose-6-phosphate. All reactions were performed at 37 °C by consecutive addition of reagents, and the assay is able to quantify 2DG in picomole per well. To confirm the reliability of this method, we have evaluated the dose-effect of insulin, GLUT4 inhibitors and insulin-sensitizing agent on 2DG uptake into 3T3-L1 cells. The results obtained with the assay for 2DG uptake in vitro in the absence or presence of insulin stimulation, were similar to those obtained by the previous radioisotopic and enzymatic methods. We have also used this assay to evaluate the effect of various reactive carbonyl and oxygen species on insulin-stimulated 2DG-uptake into adipocytes. All reactive carbonyl species tested decreased insulin-stimulated glucose uptake in a time- and dose-dependent manner without affecting basal glucose uptake in 3T3-L1 cells. 4-hydroxynonenal was found to be the most potent in the impairment of glucose uptake. This new enzymatic chemiluminescent assay is rapid and useful for measurement of 2DG uptake in insulin-responsive in cultured cells.
Sigma-Aldrich
Cytochalasin B from Drechslera dematioidea, ≥98% (HPLC), powder
Sigma-Aldrich
Cytochalasin B from Drechslera dematioidea, Ready Made Solution, 10 mg/mL in DMSO