Merck
  • A systematic study of transfection efficiency and cytotoxicity in HeLa cells using iron oxide nanoparticles prepared with organic and inorganic bases.
A systematic study of transfection efficiency and cytotoxicity in HeLa cells using iron oxide nanoparticles prepared with organic and inorganic bases.
Colloids and surfaces. B, Biointerfaces (2012-07-07)
Marilia Freitas Calmon, Aryane Tofanello de Souza, Natalia Maria Candido, Maria Irene Bartolomeu Raposo, Sebastião Taboga, Paula Rahal, Jose G Nery
ABSTRACT
Magnetic iron oxide nanoparticles (magnetite) (MNPs) were prepared using different organic and inorganic bases. Strong inorganic base (KOH) and organic bases (NH(4)OH and 1,4-diazabicyclo[2.2.2]octane (DABCO)) were used in the syntheses of the MNPs. The MNPs were characterized by X-ray diffraction (XRD), scanning electron microscope (SEM), Fourier transform infrared spectroscopy (FT-IR) and magnetization measurements. MNPs prepared with strong inorganic base yielded an average size of 100 nm, whereas the average size of the MNPs prepared with the organic bases was 150 nm. The main competitive phase for MNPs prepared with the strong inorganic and organic bases was maghemite; however, syntheses with KOH yielded a pure magnetite phase. The transfection study performed with the MNPs revealed that the highest transfection rate was obtained with the MNPs prepared with KOH (74%). The correlation between the magnetic parameters and the transfection ratio without transfection agents indicated that MNPs prepared with KOH were a better vector for possible applications of these MNPs in biomedicine. HeLa cells incubated with MNP-KOH at 10 μg/mL for 24 and 48 h exhibited a decrease in population in comparison with the control cells and it was presumably related to the toxicity of the MNPs. However, the cells incubated with MNP-KOH at 50 and 100 μg/mL presented a very small difference in the viability between the cell populations studied at 24 and 48 h. These data illustrate the viability of HeLa cells treated with MNP-KOH and suggest the potential use of these MNPs in biomedical applications.
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Ammonium hydroxide solution, puriss., 30-33% NH3 in H2O
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a. plus, ≥25% NH3 in H2O
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a., reag. ISO, reag. Ph. Eur., ~25% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Ammonium hydroxide solution, puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Dabco® 33-LV
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%