Merck
  • Hydrogen sulfide mediates hypoxic vasoconstriction through a production of mitochondrial ROS in trout gills.
Hydrogen sulfide mediates hypoxic vasoconstriction through a production of mitochondrial ROS in trout gills.
American journal of physiology. Regulatory, integrative and comparative physiology (2012-06-29)
Nini Skovgaard, Kenneth R Olson
ABSTRACT
Hypoxic pulmonary vasoconstriction (HPV) is an adaptive response that diverts pulmonary blood flow from poorly ventilated and hypoxic areas of the lung to more well-ventilated parts. This response is important for the local matching of blood perfusion to ventilation and improves pulmonary gas exchange efficiency. HPV is an ancient and highly conserved response, expressed in the respiratory organs of all vertebrates, including lungs of mammals, birds, and reptiles; amphibian skin; and fish gills. The mechanism underlying HPV and how cells sense low Po(2) remains elusive. In perfused trout gills (Oncorhynchus mykiss), acute hypoxia, as well as H(2)S, caused an initial and transient constriction of the vasculature. Inhibition of the enzymes cystathionine-β-synthase and cystathionine-γ-lyase, which blocks H(2)S production, abolished the hypoxic response. Individually blocking the four complexes in the electron transport chain abolished both the hypoxic and the H(2)S-mediated constriction. Glutathione, an antioxidant and scavenger of superoxide, attenuated the vasoconstriction in response to hypoxia and H(2)S. Furthermore, diethyldithiocarbamate, an inhibitor of superoxide dismutase, attenuated the hypoxic and H(2)S constriction. This strongly suggests that H(2)S mediates the hypoxic vasoconstriction in trout gills. H(2)S may stimulate the mitochondrial production of superoxide, which is then converted to hydrogen peroxide (H(2)O(2)). Thus, H(2)O(2) may act as the "downstream" signaling molecule in hypoxic vasoconstriction.
Sigma-Aldrich
Sodium diethyldithiocarbamate trihydrate, ACS reagent
Sigma-Aldrich
Ammonium diethyldithiocarbamate
Supelco
Sodium diethyldithiocarbamate trihydrate, ACS reagent, for the determination and separation of heavy metals, ≥97%
Sigma-Aldrich
Sodium diethyldithiocarbamate trihydrate