Merck
  • Biodegradation of aniline in an alkaline environment by a novel strain of the halophilic bacterium, Dietzia natronolimnaea JQ-AN.
Biodegradation of aniline in an alkaline environment by a novel strain of the halophilic bacterium, Dietzia natronolimnaea JQ-AN.
Bioresource technology (2012-05-23)
Qiong Jin, Zhongce Hu, Zanfang Jin, Lequan Qiu, Weihong Zhong, Zhiyan Pan
ABSTRACT
Dietzia natronolimnaea JQ-AN was isolated from industrial wastewater containing aniline. Under aerobic conditions, the JQ-AN strain degraded 87% of the aniline in a 300 mg L(-1) aniline solution after 120 h of shake flask incubation in a medium containing sodium acetate. This strain had an unusually high salinity tolerance in minimal medium (0-6% NaCl, w/v). The optimal pH for microbial growth and aniline biodegradation was pH 8.0. Two liters of simulated aniline wastewater was created in a reactor at pH 8.0 and 3% NaCl (w/v), and biodegradation of aniline was tested over 7 days at 30 °C. For the initial concentrations of 100, 300, and 500 mg L(-1), 100%, 80.5% and 72% of the aniline was degraded, respectively. Strain JQ-AN may use an ortho-cleavage pathway for dissimilation of the catechol intermediate.
Sigma-Aldrich
Sodium acetate trihydrate, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium acetate trihydrate, meets USP testing specifications
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al
Sigma-Aldrich
Sodium acetate trihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%