Merck
  • Co-occurrence of 1,4-dioxane with trichloroethylene in chlorinated solvent groundwater plumes at US Air Force installations: Fact or fiction.
Co-occurrence of 1,4-dioxane with trichloroethylene in chlorinated solvent groundwater plumes at US Air Force installations: Fact or fiction.
Integrated environmental assessment and management (2012-04-12)
Richard H Anderson, Janet K Anderson, Paul A Bower
ABSTRACT
Increasing regulatory attention to 1,4-dioxane has prompted the United States Air Force (USAF) to evaluate potential environmental liabilities, primarily associated with legacy contamination, at an enterprise scale. Although accurately quantifying environmental liability is operationally difficult given limited historic environmental monitoring data, 1,4-dioxane is a known constituent (i.e., stabilizer) of chlorinated solvents, in particular 1,1,1-trichloroethane (TCA). Evidence regarding the co-occurrence of 1,4-dioxane and trichloroethylene (TCE), however, has been heavily debated. In fact, the prevailing opinion is that 1,4-dioxane was not a constituent of past TCE formulations and, therefore, these 2 contaminants would not likely co-occur in the same groundwater plume. Because historic handling, storage, and disposal practices of chlorinated solvents have resulted in widespread groundwater contamination at USAF installations, significant potential exists for unidentified 1,4-dioxane contamination. Therefore, the objective of this investigation is to determine the extent to which 1,4-dioxane co-occurs with TCE compared to TCA, and if these chemicals are co-contaminants, whether or not there is significant correlation using available monitoring data. To accomplish these objectives, the USAF Environmental Restoration Program Information Management System (ERPIMS) was queried for all relevant records for groundwater monitoring wells (GMWs) with 1,4-dioxane, TCA, and TCE, on which both categorical and quantitative analyses were carried out. Overall, ERPIMS contained 5788 GMWs from 49 installations with records for 1,4-dioxane, TCE, and TCA analytes. 1,4-Dioxane was observed in 17.4% of the GMWs with detections for TCE and/or TCA, which accounted for 93.7% of all 1,4-dioxane detections, verifying that 1,4-dioxane is seldom found independent of chlorinated solvent contamination. Surprisingly, 64.4% of all 1,4-dioxane detections were associated with TCE independently. Given the extensive data set, these results conclusively demonstrate for the first time that 1,4-dioxane is a relatively common groundwater co-contaminant with TCE. Trend analysis demonstrated a positive log-linear relationship where median 1,4-dioxane levels increased between approximately 6% and approximately 20% of the increase in TCE levels. In conclusion, this data mining exercise suggests that 1,4-dioxane has a probability of co-occurrence of approximately 17% with either TCE and/or TCA. Given the challenges imposed by remediation of 1,4-dioxane and the pending promulgation of a federal regulatory standard, environmental project managers should use the information presented in this article for prioritization of future characterization efforts to respond to the emerging issue. Importantly, site investigations should consider 1,4-dioxane a potential co-contaminant of TCE in groundwater plumes.
Product Description
Supelco
1,1,1-Trichloroethane solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 10 mm × 8 in.
Supelco
1,4-Dioxane solution, NMR reference standard, 5 mM in benzene-d6 (99.6 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 3 mm × 8 in.
Supelco
1,4-Dioxane solution, certified reference material, 2000 μg/mL in methanol, ampule of 1 mL
Sigma-Aldrich
1,4-Dioxane solution, NMR reference standard, 10 mM in D2O ("100%", 99.96 atom % D), NMR tube size 5 mm × 7 in.
Supelco
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 5 mm × 8 in.
Supelco
1,4-Dioxane solution, NMR reference standard, 5 mM in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
1,4-Dioxane, anhydrous, 99.8%, contains <=25 ppm BHT as stabilizer
Supelco
1,4-Dioxane, analytical standard
Sigma-Aldrich
1,4-Dioxane, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D)
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
1,4-Dioxane, ReagentPlus®, ≥99%, contains ≤25 ppm BHT as stabilizer
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains ≤25 ppm BHT as stabilizer
Sigma-Aldrich
1,4-Dioxane, suitable for HPLC, ≥99.5%