Skip to Content
Merck
All Photos(1)

Key Documents

CRM48367

Supelco

1,4-Dioxane solution

certified reference material, 2000 μg/mL in methanol, ampule of 1 mL

Synonym(s):

1,4-Dioxane solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$46.80

$46.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$46.80

About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$46.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos E Agudelo-Morales et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 13(18), 4195-4201 (2012-10-24)
The quenching of pyrene and 1-methylpyrene fluorescence by nitroanilines (NAs), such as 2-, 3-, and 4-nitroaniline (2-NA, 3-NA, and 4-NA, respectively), 4-methyl-3-nitroaniline (4-M-3-NA), 2-methyl-4-nitroaniline (2-M-4-NA), and 4-methyl-3,5-dinitroaniline (4-M-3,5-DNA), are studied in toluene and 1,4-dioxane. Steady-state fluorescence data show the higher
Iñaki Galve et al.
Molecular diversity, 16(4), 639-649 (2012-10-12)
A practical protocol was developed for the synthesis of 2-arylamino substituted 4-amino-5,6-dihydropyrido[2,3-d]pyrimidin-7(8H)-ones from α,β-unsaturated esters, malononitrile, and an aryl substituted guanidine via the corresponding 3-aryl-3,4,5,6- tetrahydropyrido[2,3-d]pyrimidin-7(8H)-ones. Such compounds are formed upon treatment of 2-methoxy-6-oxo-1,4,5,6-tetrahydropyridine-3-carbonitriles with an aryl substituted guanidine in
Abdullah M Asiri et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 95, 679-684 (2012-05-18)
The absorption and fluorescence characteristics of 4-(p-N,N-dimethyl-aminophenylmethylene)-2-phenyl-5-oxazolone (DPO) have been investigated in different solvents. DPO dye exhibits a large red shift in both absorption and emission spectra as solvent polarity increases, indicating a large change in the dipole moment of
Kiyoaki Ishimoto et al.
Biomacromolecules, 13(11), 3757-3768 (2012-09-19)
For developing broader application of biobased polymers, graft copolymers and comb polymers having poly(lactic acid) (PLA) side chains have been synthesized by using a macromonomer technique. PLA macromonomers (MMm) having a methacryloyl polymerizable group with different PLA chain length with
R Dario Arrua et al.
Journal of chromatography. A, 1273, 26-33 (2013-01-01)
Rigid monolithic cryostructures were prepared in capillary format at sub-zero temperatures and used successfully in the separation of proteins by hydrophobic interaction chromatography (HIC). The polymerization mixture consisted of poly(ethyleneglycol) diacrylate (PEGDA) M(n)∼258 as the single monomer, a mixture of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service