Merck
  • Coaxial organic p-n heterojunction nanowire arrays: one-step synthesis and photoelectric properties.
Coaxial organic p-n heterojunction nanowire arrays: one-step synthesis and photoelectric properties.
Advanced materials (Deerfield Beach, Fla.) (2012-04-11)
Qiu Hong Cui, Lang Jiang, Chuang Zhang, Yong Sheng Zhao, Wenping Hu, Jiannian Yao
ABSTRACT
Organic/organic single-crystal coaxial p-n heterojunction nanowire arrays consisting of p-type copper phthalocyanine (CuPc) and n-type 5,10,15,20-tetra(4-pyridyl)-porphyrin (H(2)TPyP) are fabricated through a one-step physical vapor transport (PVT) process. Each single junction wire revealed high photodependent rectifying and sensitive photoresponsive characteristics in devices of light-controlled diodes and photoswitches. The typical photovoltaic device based on a single p-n junction exhibited a high open-circuit voltage (Voc) of 0.64 V.
Sigma-Aldrich
Copper(II) phthalocyanine, sublimed grade, Dye content 99 %
Sigma-Aldrich
Copper(II) phthalocyanine, triple-sublimed grade, >99.95% trace metals basis
Sigma-Aldrich
Copper(II) phthalocyanine, β-form, Dye content 90 %
Sigma-Aldrich
Copper(II) phthalocyanine, Dye content >99 %