Merck
  • LC/MS method using cloud point extraction for the determination of permitted and illegal food colors in liquid, semiliquid, and solid food matrixes: single-laboratory validation.
LC/MS method using cloud point extraction for the determination of permitted and illegal food colors in liquid, semiliquid, and solid food matrixes: single-laboratory validation.
Journal of AOAC International (2012-02-11)
Ebru Ates, Klaus Mittendorf, Hamide Senyuva
ABSTRACT
A cloud point extraction method is reported using LC/MS for the determination of regulated water-soluble food colors (Allura Red, Sunset Yellow, erythrosine, and tartrazine) and banned fat-soluble synthetic azo dyes (Sudan I, II, III, and IV; Red B; 7B; Black B; Red G; Metanil Yellow; and Rhodamine B). The extraction of all 14 colors was carried out with cloud point extraction using the nonionic surfactant Triton X 114. Optimized conditions for cloud point extraction were 3% Triton X 114 (w/v), 0.1 M ammonium acetate, and heating at 50 degrees C for 30 min. This approach proved effective in giving quantitative recoveries from a diverse range of food matrixes, and optimized LC gave baseline chromatographic separation for all colors including Sudan IV and Red B. Single-laboratory validation was performed with spiking into liquid matrixes (wine and homemade wine), semiliquid matrixes (sauce and homemade paprika paste), and solid matrixes (spice and homemade chili powder) using the respective blank matrixes for matrix-matched calibration. The LOQ values for water-soluble colors were in the range of 15-150 mg/kg, and for the fat-soluble colors, 0.1-1.5 mg/kg. The mean recovery values were in the range of 69.6-116.0% (except Allura Red and Sunset Yellow in wine, for which recoveries were lower). The mean RSDs for colors were in the range of 4.0-14.8%. A small survey was conducted of samples of confectionery products, dried fruits, wines, bitter sodas, juices, sauces, pastes, and spices, which demonstrated the applicability of the method to a diverse selection of real food samples. Allura Red was detected in strawberry jelly and Sunset Yellow in artificial saffron.
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
Triton X-114
Sigma-Aldrich
Triton X-165 solution, Non-ionic, CMC-0.43 mM, suitable for electrophoresis, cloud point >100 °C
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Sigma-Aldrich
IGEPAL® CA-720, average Mn ~735
Sigma-Aldrich
Triton X-114, laboratory grade
Supelco
IGEPAL® CA-630
Sigma-Aldrich
Triton X-405 solution, 70% in H2O