Merck
  • Organic-solvent extraction of model biomaterials for use in the in vitro chromosome aberration test.
Organic-solvent extraction of model biomaterials for use in the in vitro chromosome aberration test.
Journal of biomedical materials research. Part A (2007-10-18)
Atsuko Matsuoka, Yuji Haishima, Chie Hasegawa, Yoshie Matsuda, Toshie Tsuchiya
ABSTRACT
We prepared polyurethane (PU) containing 0.4% or 4% 4,4'-methylenedianiline (MDA) as model materials to investigate the effectiveness of sample preparation by organic-solvent extraction for the in vitro chromosome aberration (CA) test. MDA itself (0.4 mg/mL) was positive only in the presence of an exogenous metabolizing system (S9 mix). The culture medium extract of PU containing 4% MDA (PU/4% MDA) was negative with and without S9 mix. Methanol and acetone extracts, on the other hand, induced structural CAs without S9 mix, which we did not expect because MDA requires S9 mix for activity. On chemical analysis, however, we found that the ratio of MDA extracted by the organic solvents to that extracted by the culture medium of PU/4% MDA was about 15:1. Interestingly, oligomers consisting of poly(tetramethyleneglycol) derivatives (OTMG) were also extracted by the organic solvents. The data suggest that the induction of structural CAs in the absence of S9 mix may have been partly due to synergism of MDA and OTMG. CA tests of MDA and PTMG-1000 in combination confirmed that to be the case. Thus, organic-solvent extraction may be more effective than medium extraction in evaluating the biological safety of biomaterials. Detailed chemical analysis of extracts was performed.
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Supelco
4,4′-Diaminodiphenylmethane, analytical standard
Sigma-Aldrich
Poly(tetrahydrofuran), average Mn ~2,000, contains BHT as stabilizer
Sigma-Aldrich
Poly(tetrahydrofuran), average Mn ~2,900
Sigma-Aldrich
Poly(tetrahydrofuran), average Mn ~1,000