Skip to Content
Merck
All Photos(2)

Documents

345334

Sigma-Aldrich

Poly(tetrahydrofuran)

average Mn ~2,900

Synonym(s):

α-Hydro-ω-hydroxypoly(oxy-1,4-butanediyl), Poly(1,4-butanediol), polyTHF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2CH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 25 °C)
<1 mmHg ( 20 °C)

form

solid

mol wt

average Mn ~2,900

mp

30-43 °C

density

0.97 g/mL at 25 °C

SMILES string

OCCCCO

InChI

1S/C8H18O2/c1-3-5-6-10-8(4-2)7-9/h8-9H,3-7H2,1-2H3/t8-/m0/s1

InChI key

BJZYYSAMLOBSDY-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(tetrahydrofuran) (PTHF) is a polyether that can be prepared by the cationic polymerization of tetrahydrofuran in the presence of a variety of initiator systems.

Application

PTHF can be used in the preparation of soft segment polyurethane for a variety of applications. It can also be used in the surface modification of cellulose fibers which can further be used as reinforcing polymerix materials.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>325.4 °F - Tag open cup

Flash Point(C)

> 163 °C - Tag open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mass spectral analysis of low-temperature pyrolysis products from poly (tetrahydrofuran)
Lattimer RP
Journal of Analytical and Applied Pyrolysis, 57(1), 57-76 (2001)
Surface functionalization of cellulose by grafting oligoether chains
Bras J, et al.
Materials Chemistry and Physics, 120(2-3), 438-445 (2010)
25. Polytetrahydrofuran
Sugar Series, 13, 186-204 (2000)
Melting behavior of poly (tetrahydrofuran)-S and their blends
Tsuchiya M and Kojima T
Journal of Thermal Analysis and Calorimetry, 72(2), 651-655 (2003)
W K Loke et al.
Biomaterials, 17(22), 2163-2172 (1996-11-01)
Hybrid biomaterials have been produced by the interaction of polyurethane oligomers with both fresh and glutaraldehyde-fixed porcine pericardium. The hybrid biomaterials so formed were translucent with occasional white streaks and/or spots, had increased stiffness (to touch) but remained pliable. No

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service