Merck
  • Analysis of methanol or formic acid in body fluids by headspace solid-phase microextraction and capillary gas chromatography.
Analysis of methanol or formic acid in body fluids by headspace solid-phase microextraction and capillary gas chromatography.
Journal of chromatography. B, Biomedical sciences and applications (1999-11-26)
X P Lee, T Kumazawa, K Kondo, K Sato, O Suzuki
ABSTRACT
Methanol and its metabolite formic acid have been found extractable from human whole blood and urine by headspace solid-phase microextraction (SPME) with a Carboxen/polydimethylsiloxane fiber. The headspace SPME for formic acid was carried out after derivatization to methyl formate under acidic conditions. The determinations of both compounds were made by using acetonitrile as internal standard (IS) and capillary gas chromatography (GC) with flame ionization detection. The headspace SPME-GC gave sharp peaks for methanol, methyl formate and I.S.; and low background noises for whole blood and urine samples. Extraction efficiencies were 0.25-1.05% of methanol and 0.38-0.84% formic acid for whole blood and urine. The calibration curves for methanol and formic acid showed excellent linearity in the range of 1.56 to 800 and 1.56 to 500 microg/0.5 ml of whole blood or urine, respectively. The detection limits were 0.1-0.5 microg/0.5 ml for methanol and 0.6 microg/0.5 ml for formic acid for both body fluids. The within-day relative standard deviations in terms of extraction efficiency for both compounds in whole blood and urine samples were not greater than 9.8%. By using the established SPME method, methanol and formic acid were successfully separated and determined in rat blood after oral administration of methanol.
Product Description
Supelco
Carboxen® 569, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
ORBO 78 HBr on Carboxen® 564 specially cleaned (20/45), 400/200 mg, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Tenax® TA / Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4 in. × 3 1/2 in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, bottle of 10 g
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Sigma-Aldrich
Methyl formate, anhydrous, 99%
Supelco
Methanol, analytical standard
Supelco
Methyl formate, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Supelco
Carboxen® 1000 Reversible SPE Tube, 200 mg/mL, pk 30
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler