Skip to Content
Merck
All Photos(2)

Key Documents

28718-U

Supelco

Tenax® TA / Carboxen® 1018 Thermal Desorption Tube

greener alternative

glass, O.D. x I.D. x L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea

Synonym(s):

Tenax TA/Carbon Thermal Desorption Tube (stainless)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

Product Name

Tenax® TA / Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4 in. × 3 1/2 in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea

material

glass TD tube

Quality Level

Agency

EPA TO-1,IP-1B

feature

preconditioned

packaging

pkg of 10 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

active air sampling: suitable

O.D. × L

1/4 in. × 3 1/2 in.

matrix

Carboxen® 1018 carbon molecular sieve (CMS)
Tenax TA porous polymer

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments

greener alternative category

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.

Application

Tenax® TA / Carboxen® 1018 sorption tube may be used for the trapping and preconcentration of volatile organic compounds in breath and skin samples, followed by analysis using gas chromatography coupled with mass spectrometry and nanomaterial-based chemical sensor array.

Legal Information

Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany
Swagelok is a registered trademark of Swagelok Company
Tenax is a registered trademark of Buchem B.V.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combined volatolomics for monitoring of human body chemistry.
Broza YY, et al.
Scientific reports, 4, 4611-4611 (2014)
Sebastien Allard et al.
Journal of chromatography. A, 1238, 15-21 (2012-04-13)
Trihalomethanes are predominantly formed during disinfection of water via reactions of the oxidant with natural organic matter. Even though chlorinated and brominated trihalomethanes are the most widespread organic contaminants in drinking water, when iodide is present in raw water iodinated
Dawn M Chapman et al.
Journal of agricultural and food chemistry, 52(17), 5431-5435 (2004-08-19)
A rapid and automated solid phase microextraction (SPME) stable isotope dilution gas chromatography/mass spectrometry (GC-MS) method for 2-methoxy-3-isobutylpyrazine (MIBP) quantification in red wine was developed. Wines with 30% (w/v) NaCl and 2-methoxy-(2)H(3)-3-isobutylpyrazine internal standard were sampled with a 2 cm
Louise Urruty et al.
Journal of agricultural and food chemistry, 50(11), 3129-3136 (2002-05-16)
To provide an efficient and running analytical tool to strawberry plant breeders who have to characterize and compare the aromatic properties of new cultivars to those already known, a HS-SPME/GC-MS analysis method has been coupled with a statistical treatment method
Shih-Wei Tsai et al.
Journal of chromatography. A, 991(1), 1-11 (2003-04-22)
The solid-phase microextraction (SPME) device was used as a time-weighted average (TWA) sampler for ethylene oxide. Carboxen/polydimethylsiloxane (CAR/PDMS) fiber was used and hydrogen bromide (HBr) was loaded onto the fiber. The SPME fiber assembly was then inserted into PTFE tubing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service