Merck
  • Development of a modified DNA extraction method for pulsed-field gel electrophoresis analysis of Staphylococcus aureus and enterococci without using lysostaphin.
Development of a modified DNA extraction method for pulsed-field gel electrophoresis analysis of Staphylococcus aureus and enterococci without using lysostaphin.
Journal of microbiological methods (2010-11-17)
Mohammad Mehdi Feizabadi, Arash Ghodousi, Bizhan Nomanpour, Maryam Omrani, Fereshteh Shahcheraghi
ABSTRACT
A modified pulsed-field gel electrophoresis (PFGE) protocol was developed and applied to clinical isolates of Staphylococcus aureus and enterococci to reduce the cost of using lysostaphin. This protocol reduces the expenses of PFGE typing of S. aureus and enterococci as it removes the use of lysostaphin during the spheroplast formation from these bacteria.
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, lyophilized powder, Protein 50-70 % by biuret, ≥500 units/mg protein
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, BioUltra, ≥97% (SDS-PAGE), Protein 40-60 % by biuret, ≥2,000 units/mg protein
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, aseptically filled
Sigma-Aldrich
Lysostaphin from Staphylococcus simulans, recombinant, expressed in E. coli, lyophilized powder