Merck
  • [Study on the extraction process of total flavonoids from Psoralea corylifolia by enzyme-assisted technique].
[Study on the extraction process of total flavonoids from Psoralea corylifolia by enzyme-assisted technique].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2012-12-21)
You-Shao Yan, Hua-Wei Liao, Li-Bing Guo
ABSTRACT
To study the optimum extraction process of total flavonoids from Psoralea coryl folia by cellulose-assisted technique. Based on single-factor experiments, the effects of pH value, temperature of enzymatic hydrolysis, time of enzymatic hydrolysis and enzyme amount on the extraction yields of total flavonoids from Psoralea corylifolia were studied by response surface methodology. The optimum enzyme-assisted extraction process was: pH value 4.9, temperature 46 degrees C, time 150 min and enzyme amount 7.2 mg/g,under this condition,the relative error of the observed and predicted values was 1.16%. The optimum enzyme-assisted extraction process is simple and feasible, the extraction rate of total flavonoids increases by 28% compared with ultrasonic extraction, so it can be used to extract total flavonoids from Psoralea corylifolia.
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, ≥0.3 units/mg solid
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, off-white, ~0.8 U/mg
Sigma-Aldrich
Cellulase from Aspergillus sp., aqueous solution
Sigma-Aldrich
Cellulase from Trichoderma reesei, aqueous solution, ≥700 units/g
Sigma-Aldrich
Cellulase from Trichoderma reesei ATCC 26921, lyophilized powder, ≥1 unit/mg solid
Sigma-Aldrich
Cellulase from Trichoderma sp., BioReagent, suitable for plant cell culture, 3-10 units/mg solid
Sigma-Aldrich
Cellulase from Trichoderma sp., powder, ≥5,000 units/g solid