Merck
[Screening of two straw-cellulose degrading actinomycetes].
Wei sheng wu xue bao = Acta microbiologica Sinica (2012-12-15)
Wenjie Gu, Fabao Zhang, Peizhi Xu, Kaizhi Xie, Shuanhu Tang
ABSTRACT
The aim of this study was to screen microorganisms that could degrade rice straw. We used selective medium to screen strains and determined straw fracture tension strength, weight loss, lignocellulose decomposition rate and extracellular enzyme activity as re-screening methods after 10 days shake flask culture. We isolated two antinomycetes (A3 and A6), the highest cellulose enzyme activity of holoenzyme, beta3-Glucosidase, endonuclease and exonclease for A3 were 12.84, 6.23, 24.56 and 14.00 U/mL, and for A6 12.85, 6.53, 17.80 and 18.80 U/mL. The hemicelluloses enzyme activity was 83.05 for A3 and 52.98 U/mL for A6. Both strains belonged to Streptomyces. With 10 days' treatment, inoculated straws showed a decrease of straw fracture tension strength by 62.67% (A3) and 66.67% (A6), while weight loss of straw was 31.50% (A3) and 35.83% (A6). A3's decomposition rate of cellulose, hemicellulose and lignin was 38.73% , 33.16% and 20.68% , and 47.69% , 28.64% and 22.59% for A6. Antinomycetes A3 and A6 could degrad cellulose, hemicellulose and lignin.
Sigma-Aldrich
Cellulase from Aspergillus sp., aqueous solution
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, ≥0.3 units/mg solid
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, off-white, ~0.8 U/mg
Sigma-Aldrich
Cellulase from Trichoderma reesei, aqueous solution, ≥700 units/g
Sigma-Aldrich
Cellulase from Trichoderma sp., BioReagent, suitable for plant cell culture, 3-10 units/mg solid
Sigma-Aldrich
Cellulase from Trichoderma reesei ATCC 26921, lyophilized powder, ≥1 unit/mg solid
Sigma-Aldrich
Cellulase from Trichoderma sp., powder, ≥5,000 units/g solid