Merck
  • The transcription factor Stat-1 is essential for Schwann cell differentiation, myelination and myelin sheath regeneration.
The transcription factor Stat-1 is essential for Schwann cell differentiation, myelination and myelin sheath regeneration.
Molecular medicine (Cambridge, Mass.) (2023-06-27)
Jinghui Xu, Bin Zhang, Jieyi Cai, Qianqian Peng, Junxia Hu, Parizat Askar, Jianghong Shangguan, Wenfeng Su, Changlai Zhu, Hualin Sun, Songlin Zhou, Gang Chen, Xiaoming Yang, Yun Gu
ABSTRACT
Myelin sheath is a crucial accessory to the functional nerve-fiber unit, its disruption or loss can lead to axonal degeneration and subsequent neurodegenerative diseases (NDs). Notwithstanding of substantial progress in possible molecular mechanisms underlying myelination, there is no therapeutics that prevent demyelination in NDs. Therefore, it is crucial to seek for potential intervention targets. Here, we focused on the transcriptional factor, signal transducer and activator of transcription 1 (Stat1), to explore its effects on myelination and its potential as a drug target. By analyzing the transcriptome data obtained from Schwann cells (SCs) at different stages of myelination, it was found that Stat1 might be involved in myelination. To test this, we used the following experiments: (1) In vivo, the effect of Stat1 on remyelination was observed in an in vivo myelination mode with Stat1 knockdown in sciatic nerves or specific knockdown in SCs. (2) In vitro, the RNA interference combined with cell proliferation assay, scratch assay, SC aggregate sphere migration assay, and a SC differentiation model, were used to assess the effects of Stat1 on SC proliferation, migration and differentiation. Chromatin immunoprecipitation sequencing (ChIP-Seq), RNA-Seq, ChIP-qPCR and luciferase activity reporter assay were performed to investigate the possible mechanisms of Stat1 regulating myelination. Stat1 is important for myelination. Stat1 knockdown in nerve or in SCs reduces the axonal remyelination in the injured sciatic nerve of rats. Deletion of Stat1 in SCs blocks SC differentiation thereby inhibiting the myelination program. Stat1 interacts with the promoter of Rab11-family interacting protein 1 (Rab11fip1) to initiate SC differentiation. Our findings demonstrate that Stat1 regulates SC differentiation to control myelinogenic programs and repair, uncover a novel function of Stat1, providing a candidate molecule for clinical intervention in demyelinating diseases.
Sigma-Aldrich
Monoclonal Anti-S-100 (β-Subunit) antibody produced in mouse, clone SH-B1, ascites fluid
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥96% (HPLC), powder
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
5-Fluoro-2′-deoxyuridine, thymidylate synthase inhibitor