Merck
  • Expansion of ventral foregut is linked to changes in the enhancer landscape for organ-specific differentiation.
Expansion of ventral foregut is linked to changes in the enhancer landscape for organ-specific differentiation.
Nature cell biology (2023-01-24)
Yan Fung Wong, Yatendra Kumar, Martin Proks, Jose Alejandro Romero Herrera, Michaela Mrugala Rothová, Rita S Monteiro, Sara Pozzi, Rachel E Jennings, Neil A Hanley, Wendy A Bickmore, Joshua M Brickman
ABSTRACT
Cell proliferation is fundamental for almost all stages of development and differentiation that require an increase in cell number. Although cell cycle phase has been associated with differentiation, the actual process of proliferation has not been considered as having a specific role. Here we exploit human embryonic stem cell-derived endodermal progenitors that we find are an in vitro model for the ventral foregut. These cells exhibit expansion-dependent increases in differentiation efficiency to pancreatic progenitors that are linked to organ-specific enhancer priming at the level of chromatin accessibility and the decommissioning of lineage-inappropriate enhancers. Our findings suggest that cell proliferation in embryonic development is about more than tissue expansion; it is required to ensure equilibration of gene regulatory networks allowing cells to become primed for future differentiation. Expansion of lineage-specific intermediates may therefore be an important step in achieving high-fidelity in vitro differentiation.
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Sigma-Aldrich
SANT-1, ≥98% (HPLC), powder
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit