Skip to Content
Merck
All Photos(3)

Documents

S4572

Sigma-Aldrich

SANT-1

≥98% (HPLC), powder, SHH antagonist

Synonym(s):

(4-Benzyl-piperazin-1-yl)-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-ylmethylene)-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27N5
CAS Number:
Molecular Weight:
373.49
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

product name

SANT-1, ≥98% (HPLC), powder

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

Cc1nn(-c2ccccc2)c(C)c1\C=N\N3CCN(CC3)Cc4ccccc4

InChI

1S/C23H27N5/c1-19-23(20(2)28(25-19)22-11-7-4-8-12-22)17-24-27-15-13-26(14-16-27)18-21-9-5-3-6-10-21/h3-12,17H,13-16,18H2,1-2H3/b24-17+

InChI key

FOORCIAZMIWALX-JJIBRWJFSA-N

Application

  • SANT-1 has been used: to treat cells and study the effect of SANT1-mediated inhibition on hedgehog signaling
  • in the generation of functional human pancreatic β cells in vitro
  • to treat of Anaplastic lymphoma kinase (ALK)-positive anaplastic large cell lymphoma cells2 and chronic lymphocytic leukemia cells3 with SANT-1 induced cell death

Biochem/physiol Actions

SANT-1 is a potent sonic hedgehog pathway (Shh) antagonist that directly inhibits by binding to the smoothened (Smo) receptor. SANT-1 inhibits wild type and oncogenic Smo with equal potency.
SANT-1 is known to induce apoptosis via Ras/NF-κB pathway in glioblastoma cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deobrat Dixit et al.
Cancer letters, 336(2), 347-358 (2013-04-04)
Since Shh pathway effector, Gli1, is overexpressed in gliomas, we investigated the effect of novel Shh inhibitor SANT-1 on glioma cell viability. Though SANT-1 failed to induce apoptosis, it reduced proliferation of glioma stem-like cells. Apart from canonical Shh cascade
Felicia W Pagliuca et al.
Cell, 159(2), 428-439 (2014-10-11)
The generation of insulin-producing pancreatic β cells from stem cells in vitro would provide an unprecedented cell source for drug discovery and cell transplantation therapy in diabetes. However, insulin-producing cells previously generated from human pluripotent stem cells (hPSC) lack many
P Desch et al.
Oncogene, 29(35), 4885-4895 (2010-07-07)
The Hedgehog (Hh) pathway regulates cell proliferation and survival and contributes to tumorigenesis. We investigated the expression and function of this pathway in B-cell chronic lymphocytic leukemia (CLL) cells and in healthy B lymphocytes. Profiling of cognate Hh pathway members
Shuhei Konagaya et al.
Scientific reports, 9(1), 640-640 (2019-01-27)
Large numbers of hormone-releasing cells, approximately 109 endocrine cells, are required to treat type I diabetes patients by cell transplantation. The SOX9-positive pancreatic epithelium proliferates extensively during the early stages of pancreatic development. SOX9-positive pancreatic epithelium is thought to be
Bushra Memon et al.
Stem cell research & therapy, 9(1), 15-15 (2018-01-25)
Pancreatic progenitors (PPs) co-expressing the two transcription factors (TFs) PDX1 and NKX6.1 are recognized as the indispensable precursors of functional pancreatic β cells. Here, we aimed to establish an efficient protocol for maximizing generation of PDX1 In order to enhance

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service