Merck
  • Disruption of pancreatic stellate cell myofibroblast phenotype promotes pancreatic tumor invasion.
Disruption of pancreatic stellate cell myofibroblast phenotype promotes pancreatic tumor invasion.
Cell reports (2022-01-27)
Elizabeth R Murray, Shinelle Menezes, Jack C Henry, Josie L Williams, Lorena Alba-Castellón, Priththivika Baskaran, Ivan Quétier, Ami Desai, Jacqueline J T Marshall, Ian Rosewell, Marianthi Tatari, Vinothini Rajeeve, Faraz Khan, Jun Wang, Panoraia Kotantaki, Eleanor J Tyler, Namrata Singh, Claire S Reader, Edward P Carter, Kairbaan Hodivala-Dilke, Richard P Grose, Hemant M Kocher, Nuria Gavara, Oliver Pearce, Pedro Cutillas, John F Marshall, Angus J M Cameron
ABSTRACT
In pancreatic ductal adenocarcinoma (PDAC), differentiation of pancreatic stellate cells (PSCs) into myofibroblast-like cancer-associated fibroblasts (CAFs) can both promote and suppress tumor progression. Here, we show that the Rho effector protein kinase N2 (PKN2) is critical for PSC myofibroblast differentiation. Loss of PKN2 is associated with reduced PSC proliferation, contractility, and alpha-smooth muscle actin (α-SMA) stress fibers. In spheroid co-cultures with PDAC cells, loss of PKN2 prevents PSC invasion but, counter-intuitively, promotes invasive cancer cell outgrowth. PKN2 deletion induces a myofibroblast to inflammatory CAF switch in the PSC matrisome signature both in vitro and in vivo. Further, deletion of PKN2 in the pancreatic stroma induces more locally invasive, orthotopic pancreatic tumors. Finally, we demonstrate that a PKN2KO matrisome signature predicts poor outcome in pancreatic and other solid human cancers. Our data indicate that suppressing PSC myofibroblast function can limit important stromal tumor-suppressive mechanisms, while promoting a switch to a cancer-supporting CAF phenotype.
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Monoclonal Anti-Desmin antibody produced in mouse, clone DE-U-10, ascites fluid
Supelco
4-Hydroxytamoxifen, analytical standard, (E) and (Z) isomers (50:50)