Merck
Chaperone-mediated autophagy.
Methods in molecular biology (Clifton, N.J.) (2008-04-22)
S Kaushik, A M Cuervo
ABSTRACT
Chaperone-mediated autophagy (CMA) is the only type of autophagy in mammalian cells able to selectively degrade cytosolic proteins in lysosomes. CMA is maximally activated in response to stressors such as prolonged starvation, exposure to toxic compounds, or oxidative stress. We have found that CMA activity decreases in aging and in some age-related disorders such as Parkinson's disease. Impaired CMA under these conditions may be responsible for the accumulation of damaged proteins inside cells and for their higher vulnerability to stressors. In contrast to other forms of autophagy, where substrates are engulfed or sequestered along with other cytosolic components, CMA substrates are translocated one-by-one across the lysosomal membrane. Changes in the levels/activity of the lysosomal components required for substrate translocation can be used to stimulate CMA activity. However, the most unequivocal method to measure CMA is by directly tracking the translocation of substrate proteins into isolated lysosomes.
Sigma-Aldrich
Sarcosine, 98%
Sigma-Aldrich
Creatine Phosphokinase from bovine heart, Type III, salt-free, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Sephadex® G-100, DNA grade, BioReagent, Molecular Biology, medium
Millipore
Glutathione−Agarose, lyophilized powder
Millipore
Glutathione−Agarose, set of 3 pre-packed columns (2.5 ml each), (1:1 suspension in a 0.5 M NaCl + 20% ethanol solution)
Sigma-Aldrich
Sephadex® G-100, Superfine
Sigma-Aldrich
Sephadex® G-100, Medium
Sigma-Aldrich
Sarcosine, BioXtra
Sigma-Aldrich
Creatine Phosphokinase from rabbit muscle, Type I, salt-free, lyophilized powder, ≥150 units/mg protein
Sigma-Aldrich
Lysozyme human, Lysobac, recombinant, expressed in rice, lyophilized powder, ≥100,000 units/mg protein
Sigma-Aldrich
Sarcosine, crystallized, ≥98.0% (T)
Sigma-Aldrich
Thrombin human, BioUltra, recombinant, expressed in HEK 293 cells, aqueous solution, ≥95% (SDS-PAGE)
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Sigma-Aldrich
Chymostatin, microbial