Skip to Content
Merck
All Photos(1)

Key Documents

G3907

Millipore

Glutathione−Agarose

set of 3 pre-packed columns (2.5 ml each), (1:1 suspension in a 0.5 M NaCl + 20% ethanol solution)

Synonym(s):

GSH-agarose, S-linked glutathione agarose

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
$763.00

$763.00


Estimated to ship onMay 30, 2025



Select a Size

Change View
1 SET
$763.00

About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

$763.00


Estimated to ship onMay 30, 2025


Quality Level

form

(1:1 suspension in a 0.5 M NaCl + 20% ethanol solution)

analyte chemical class(es)

proteins (GST)

packaging

set of 3 pre-packed columns (2.5 ml each)

technique(s)

immunoprecipitation (IP): suitable
protein purification: suitable

matrix

cross-linked 4% beaded agarose

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

The resin in these columns consists of glutathione attached through the sulfur to epoxy activated, 4% cross-linked beaded agarose resulting in a 12 atom spacer. [1]

Application

Affinity chromatography using glutathione-agarose permits rapid, mild, non-denaturing and highly selective purification of glutathione binding enzymes such as glutathione-S-transferase, glutathione peroxidase, and glyoxalase I.[2]
The product was used in the design, synthesis and biological evaluation of new tryptamine and tetrahydro-β-carboline-based selective inhibitors of CDK4.[3] It was also used to study Fascaplysin-inspired diindolyls as selective inhibitors of CDK4/cyclin D1.[4]
Affinity chromatography using glutathione-agarose permits rapid, mild, non-denaturing and highly selective purification of proteins containing glutathione binding sequences, such as Glutathione S-Transferase (GST), glutathione peroxidase and glyoxalase I.

Linkage

Suspension of Product No. G 4510

Physical form

1:1 suspension in a 0.5 M NaCl + 20% ethanol solutionl

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

180.1 °F

Flash Point(C)

82.3 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carine Aubry et al.
Organic & biomolecular chemistry, 4(5), 787-801 (2006-02-24)
We present the design, synthesis, and biological activity of three classes of tryptamine derivatives, which are non-planar analogues of the toxic anti-cancer agent fascaplysin. We show these compounds to be selective inhibitors of CDK4 over CDK2, the most active compound
Diane M Kanter et al.
Nucleic acids research, 39(7), 2580-2592 (2010-11-27)
Sld2 is essential for the initiation of DNA replication, but the mechanism underlying its role in replication is not fully understood. The S-phase cyclin dependent kinase (S-CDK) triggers the association of Sld2 with Dpb11, and a phosphomimetic mutation of Sld2
F Toribio et al.
Journal of chromatography. B, Biomedical applications, 684(1-2), 77-97 (1996-09-20)
The different preparative techniques and related analytical methods used for purification of glutathione peroxidase, glutathione transferase and glutathione reductase, described in papers published in the last ten years, have been reviewed in this article. Among the different purification techniques, chromatography
Dustin L Johnson et al.
Journal of bacteriology, 190(8), 2972-2980 (2008-02-19)
Chlamydophila pneumoniae is a gram-negative obligate intracellular bacterial pathogen that causes pneumonia and bronchitis and may contribute to atherosclerosis. The developmental cycle of C. pneumoniae includes a morphological transition from an infectious extracellular elementary body (EB) to a noninfectious intracellular
Paul R Jenkins et al.
Bioorganic & medicinal chemistry, 16(16), 7728-7739 (2008-07-25)
We present the design, synthesis and biological activity of a library of substituted (biphenylcarbonyl)-tryptamine and (biphenylcarbonyl)-tetrahydro-beta-carboline compounds related to the natural product fascaplysin, as novel inhibitors of CDK4/cyclin D1. We show all these molecules, prepared using the Suzuki-Miyaura reaction, being

Related Content

Investigate in vitro protein-protein interactions with pull-down assays, utilizing affinity, GST pull-down, TAP, and co-immunoprecipitation methods.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service