Merck
  • GABA-receptive microglia selectively sculpt developing inhibitory circuits.
GABA-receptive microglia selectively sculpt developing inhibitory circuits.
Cell (2021-07-08)
Emilia Favuzzi, Shuhan Huang, Giuseppe A Saldi, Loïc Binan, Leena A Ibrahim, Marian Fernández-Otero, Yuqing Cao, Ayman Zeine, Adwoa Sefah, Karen Zheng, Qing Xu, Elizaveta Khlestova, Samouil L Farhi, Richard Bonneau, Sandeep Robert Datta, Beth Stevens, Gord Fishell
ABSTRACT
Microglia, the resident immune cells of the brain, have emerged as crucial regulators of synaptic refinement and brain wiring. However, whether the remodeling of distinct synapse types during development is mediated by specialized microglia is unknown. Here, we show that GABA-receptive microglia selectively interact with inhibitory cortical synapses during a critical window of mouse postnatal development. GABA initiates a transcriptional synapse remodeling program within these specialized microglia, which in turn sculpt inhibitory connectivity without impacting excitatory synapses. Ablation of GABAB receptors within microglia impairs this process and leads to behavioral abnormalities. These findings demonstrate that brain wiring relies on the selective communication between matched neuronal and glial cell types.
Sigma-Aldrich
DAPI, for nucleic acid staining
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-NeuN Antibody (rabbit), from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-NeuN purified Antibody, from guinea pig, purified by affinity chromatography
Sigma-Aldrich
Anti-Glutamate Decarboxylase Antibody, 65 kDa isoform, clone GAD-6, clone GAD-6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-GABA B Receptor R2 Antibody, serum, from guinea pig