Skip to Content
Merck
All Photos(1)

Documents

AB2255

Sigma-Aldrich

Anti-GABA B Receptor R2 Antibody

serum, from guinea pig

Synonym(s):

G protein-coupled receptor 51, G-protein coupled receptor 51, GABA-B receptor 2, gamma-aminobutyric acid (GABA) B receptor, 2, gamma-aminobutyric acid B receptor, 2, GABA-B-R2, GABA(B)R2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

guinea pig

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rhesus macaque, rat

species reactivity (predicted by homology)

canine (based on 100% sequence homology), bovine (based on 100% sequence homology), human (based on 100% sequence homology), horse (based on 100% sequence homology), rhesus monkey (based on 100% sequence homology), chimpanzee (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GABBR2(9568)

General description

Gamma-aminobutyric acid (GABA) is the main inhibitory neurotransmitter in the mammalian central nervous system. GABA exerts its effects through ionotropic [GABA(A/C)] receptors, to produce fast synaptic inhibition, and metabotropic [GABA(B)] receptors, to produce slow, prolonged inhibitory signals. The GABA(B) receptor consists of a heterodimer of two related 7-transmembrane receptors, GABA(B) receptor 1 and GABA(B) receptor 2. The GABA(B) receptor 1 gene is mapped to chromosome 6p21.3 within the HLA class I region close to the HLA-F gene. Susceptibility loci for multiple sclerosis, epilepsy, and schizophrenia have also been mapped in this region. Alternative splicing of this gene generates 4 transcript variants.

Specificity

This antibody recognizes the C-terminus of rat GABA-B-R2.

Immunogen

Bovine thyroglobulin-conjugated linear peptide at the C-terminus of rat GABA-B-R2.

Application

Anti-GABA B Receptor R2 Antibody is an antibody against GABA B Receptor R2 for use in IH & WB.

Quality

Evaluated by Immunohistochemistry in Purkinje cells in rat mid-brain region.

Immunohistochemistry Analysis: A 1:300 dilution of this antibody detected GABA-B-R2 in Purkinje cells in rat mid-brain region.

Target description

Although a band is observed around the calculated molecular weight of 105 kDa, Western Blotting is not a recommened application for this antibody because the band is weak in comparison to other non-specific bands present.

Linkage

Replaces: AB5394

Analysis Note

Control
Purkinje cells in rat mid-brain region

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yupeng Miao et al.
Nature communications, 12(1), 4518-4518 (2021-07-28)
Multiplexed optical imaging provides holistic visualization on a vast number of molecular targets, which has become increasingly essential for understanding complex biological processes and interactions. Vibrational microscopy has great potential owing to the sharp linewidth of vibrational spectra. In 2017
Liang Zhang et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 41(6), 1467-1476 (2015-10-27)
Cortical network hyper-excitability is a common phenotype in mouse models lacking the transcriptional regulator methyl-CPG-binding protein 2 (MeCP2). Here, we implicate enhanced GABAB receptor activity stemming from diminished cortical expression of the GABA transporter GAT-1 in the genesis of this
Chuanyin Hu et al.
Molecular medicine reports, 15(2), 975-980 (2016-12-31)
Chemotherapeutic drugs commonly induce peripheral neuropathic pain, which limit their clinic use. In the present study, the effect of fucoidan on the development of vincristine‑induced neuropathic pain was evaluated and the underlying mechanism was examined. A neuropathy model was established
Khaled Zemoura et al.
The Journal of biological chemistry, 291(41), 21682-21693 (2016-08-31)
GABAB receptors are heterodimeric G protein-coupled receptors, which control neuronal excitability by mediating prolonged inhibition. The magnitude of GABAB receptor-mediated inhibition essentially depends on the amount of receptors in the plasma membrane. However, the factors regulating cell surface expression of
Khaled Zemoura et al.
Molecular neurobiology, 56(2), 1293-1309 (2018-06-09)
The G protein-coupled GABAB receptors, constituted from GABAB1 and GABAB2 subunits, are important regulators of neuronal excitability by mediating long-lasting inhibition. One factor that determines receptor availability and thereby the strength of inhibition is regulated protein degradation. GABAB receptors are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service